Cole-Parmer® Essentials Clean Wipes, Pre-wetted
Strong, nonwoven cloth is soft and lint-free – will not scratch surfaces
  • No more struggling with solutions and separate towels—ready to use
  • Individually sealed containers for use in cleanrooms
Premoistened wipes are available either with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reagent-grade deionized water, or 100% pure reagent-grade deionized water. Individually dispensed wipes are handy and ready for instant use. Snap-on sealed top eliminates contamination from dust or dirt. Each canister is provided in shrink-wrap or a sealed bag to maintain cleanliness for use in clean rooms. Wipes are 6 x 7.5 inches and packaged 100 per polyethylene canister.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Description
Availability Pricing
COMPARE
7 1/2 Polypropylene, Wood Pulp, Viscose Blend Clean Wipes, 70% Alcohol (IPA)/30% DI Water; 100 Wipes/Canister
Call For Price
COMPARE
7 1/2 Polypropylene, Wood Pulp, Viscose Blend Clean-Wipes, 100% DI Water: 100 Wipes/Canister