This product is now discontinued, View Related Products
Cole-Parmer® Essentials Class B PMP Volumetric Flasks
View samples easily
  • Volumetric flasks are made of highly transparent PMP
  • Autoclavable
These Cole-Parmer® Class B volumetric flasks are made of durable, transparent polymethylpentene (PMP) and are supplied with individual class B calibrated ring markings. These flasks can withstand temperatures to 350°F(180°C).
6,100.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL)
Availability Pricing
COMPARE
25
Call For Price
COMPARE
50
Call For Price
COMPARE
100
Call For Price
COMPARE
250
Call For Price
COMPARE
500
₹6,100.00INR / Each
COMPARE
1000