This product is now discontinued, View Related Products
Cole-Parmer Class A Glass Media Bottles
Affordable borosilicate bottles are ideal for your storage, mixing, and sampling needs!
  • Quick identification with large marking spot and graduations
  • Wide-mouth opening for easy pouring
  • Linerless, blue polypropylene cap and pouring ring
  • Withstands short-term exposure to 752°F (400°C) and long-term exposure to 445°F (230°C)
  • Sturdy borosilicate Class A glass meets ASTM E438, Type 1, Class A requirements
  • Autoclavable
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
100 25 Glass
Call For Price
COMPARE
250 50 Glass
Call For Price
COMPARE
500 100 Glass
Call For Price
COMPARE
1000 100 Glass
Call For Price
COMPARE
2000 100 Glass