Cole-Parmer® Centrifuge Tubes
Safely withstands temperatures down to -80 °C
  • Disposable polypropylene tubes are autoclavable to 250 °F (121 °C)
  • Can be safely stored in freezers down to -112 °F (-80 °C)
  • Caps with recessed lid and sealing ring prevent leaking
Easy-to-read black graduations offer accurate reading to ±2%; the 15-mL tubes feature increments of 0.5 mL while the 50-mL tubes have increments of 2.5 mL. Frosted writing surface allows for permanent identification and labeling. Tubes are offered in either non-sterilized or sterilized versions, are nonpyrogenic, and certified RNase-, DNase- and DNA-free.
Available in cardboard racks or in resealable zip-top bags.
31,700.00 - ₹54,300.00INR / Case of 500
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Sterile Tube Capacity (mL)
Availability Pricing
COMPARE
12000 Yes 15
₹36,600.00INR / Case of 500
COMPARE
12000 Yes 15
₹41,900.00INR / Case of 500
COMPARE
15000 Yes 50
₹47,500.00INR / Case of 500
COMPARE
15000 Yes 50
₹54,300.00INR / Case of 500
COMPARE
12000 No 15
₹31,700.00INR / Case of 500
COMPARE
15000 No 50
₹38,600.00INR / Case of 500
COMPARE
15000 No 50
₹31,700.00INR / Case of 500
COMPARE
15000 No 15
₹31,700.00INR / Case of 500
COMPARE
15000 No 50
₹38,600.00INR / Case of 500
COMPARE
15000 No 50
₹45,800.00INR / Case of 500
Showing 1- 10 of 10