Cole-Parmer® Sterile Cell and Tissue Culture Well Plates
Extremely smooth surface and raised well rims reduce evaporation and maximize results
  • Gamma sterilized and nonpyrogenic
  • Individually packaged in peel-to-open blister pack
  • Printed lot numbers on both plate and packaging for effortless traceability
  • Optically clear plates for easy visualization of cell growth
Use these sterile well plates to serve your various cell and tissue culture needs. With individual condensation rings and a one-way, tight-fitting lid, they reduce cross-contamination and help prevent evaporation. An alphanumeric notation on each well aids in simple identification. Transparent polystyrene construction allows for easy visualization of cell growth and facilitates observation under a microscope.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment for optimal quality, these cell and tissue culture well plates are individually packaged in peel-to-open blister packs, sterilized by irradiation (SAL 10-6), and are DNase/RNase free, nonpyrogenic, and noncytotoxic. They conform to USP Class VI and are quality traceable with the lot number on both the plate and packaging.
Plates with a treated surface are suitable for common adherent cell lines due to the excellent hydrophilicity of the surface while untreated plates are suitable for suspension cell culture. Compatible with most multi-well plate instruments and equipment.
14,900.00 - ₹26,000.00INR / Case of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Culture Area (cm²) Number of Wells Surface Treatment
Availability Pricing
COMPARE
9.6 6 Yes
₹24,200.00INR / Case of 100
COMPARE
1.93 24 Yes
₹26,000.00INR / Case of 100
COMPARE
0.33 96 No
Call For Price
COMPARE
0.33 96 Yes
₹24,200.00INR / Case of 100
COMPARE
6
₹14,900.00INR / Case of 1
COMPARE
12
₹16,600.00INR / Case of 1
COMPARE
24
₹16,600.00INR / Case of 1
COMPARE
96
₹14,900.00INR / Case of 1
Showing 1- 8 of 8