Cole-Parmer Carboy Vent Filters
Ideal for hydrophobic venting and solvent filtration applications
  • Complies with USP Class VI standards
The non-sterile vent filters feature a polypropylene housing with a 0.2 µm hydrophobic PTFE membrane—ideal for maintaining purified water storage in carboys or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip™ carboys. They can withstand up to 10 autoclave cycles, feature a stepped hose barb, and are available in 40 mm gamma safe or 50 mm autoclave only membrane sizes.
117,000.00INR / Pkg of 25
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
50 0.2 Carboy Vent Filter, 50 mm, 0.2 um, Gamma Safe; 25/Cs
₹117,000.00INR / Pkg of 25