This product is now discontinued, View Related Products
Cole-Parmer Carboy Solvent Waste Systems, HDPE, 5 L
Protect workers from inhaling dangerous fumes while collecting HPLC waste
  • Capture potentially dangerous solvent vapors
  • Safely collect waste with the high-density polyethylene (HDPE) carboy which features superior chemical compatibility
  • No need to remove tubing from the open cap to install a closed cap
This heavy-duty solvent waste system is designed to collect liquid waste disposal from one or more HPLC systems while a carbon exhaust filter captures potentially harmful vapors emitted. The closed system helps eliminate spills and provides a safe and efficient way of containing, transporting, and disposing of aggressive organic solvents.
The polypropylene cap (PP) offers a wide range of tubing port configurations. The 5-L carboys come with 83 mm caps.
Call For Price
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Cap / Stopper Size Material Description
Availability Pricing
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 6 x 1/16\" OD Tubing, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 6 x 1/8\" OD Tubing, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/8\", 3 x 1/4\" OD Tube Fittings, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 6 x 1/8\" OD Tubing, 1 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/8\", 3 x 1/4\" OD Tubing, 1 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/16\", 3 x 1/4\" OD Tube Fittings, 1 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/8\", 4 x 1/4\" OD Tubing, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/16\", 3 x 1/4\" OD Tube Fittings, 1 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/16\", 1 x 1/4\" OD Tube Fittings, 3 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/8\", 1 x 1/4\" OD Tube Fittings, 3 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/8\" OD Tube Fittings, 4 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/16\" OD Tube Fittings, 4 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 6 x 1/16\" OD Tube Fittings, 1 x 1/4\" or 3/8\" Hose Barb, Exhaust Filter; 5 L
Call For Price
COMPARE
83 mm High Density Polyethyene (HDPE) Carboy Solvent Waste System, 83 mm Cap, 4 x 1/16\", 4 x 1/4\" OD Tube Fittings, Exhaust Filter; 5 L
Call For Price
Showing 1- 14 of 14