This product is now discontinued, View Related Products
Cole-Parmer® Essentials Carboy Fittings and Spigot
Use these fittings and spigot with carboys to easily dispense liquid
  • Made of autoclavable polypropylene
  • Feature a 1-1/8"-12 UNF-2B thread
Fittings are available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes for convenient connection to tubing. Use the 3/4" sanitary connection fitting to make carboys function as a reservoir to a larger system. Spigot has a hose barb to accept 5/8" to 3/4" ID tubing and a standard internal luer taper to fit a male barb adapter. The internal thread has a hard stop to prevent over-tightening which may damage the seal. Use the large lever to control two flow rates—continuous locked flow or faster manual flow.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Connection Type Description
Availability Pricing
COMPARE
5/8\" to 3/4\" hose barb Carboy spigot, 2/pk
Call For Price
COMPARE
5/8\" to 3/4\" hose barb Carboy spigot, 1/pk
Call For Price
COMPARE
Closed cap Carboy fitting, 1/pk
Call For Price
COMPARE
1/4\" hose barb Carboy fitting, 1/pk
Call For Price
COMPARE
3/8\" hose barb Carboy fitting, 1/pk
Call For Price
COMPARE
1/2\" hose barb Carboy fitting, 1/pk
Call For Price
COMPARE
3/4\" hose barb Carboy fitting, 1/pk
Call For Price
COMPARE
3/4\" sanitary fitting Carboy fitting, 1/pk
Call For Price
Showing 1- 8 of 8