Cole-Parmer Calibrated Specific Gravity ASTM Hydrometers
Easily measure specific gravity of petroleum products and heavy liquids
  • Designed to conform to the accuracy requirements of ASTM
  • Temperature standard is 60°F (15.6°C)
  • Individually serialized for easy identification
The ASTM precision hydrometers are designed to measure specific gravity of petroleum products and heavy liquids in various ranges from 0.650 to 1.500 with 0.0005 divisions. Available in various calibrated models, these lead free hydrometers are ideal for the food and beverage, chemical, pharmacology, transportation and refrigeration industries.
136,000.00 - ₹160,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
ASTM 83H Precision, Individually Calibrated 0.700/0.750 Specific Gravity Hydrometer
₹136,000.00INR / Each
COMPARE
ASTM 84H Precision, Individually Calibrated 0.750/0.800 Specific Gravity Hydrometer
₹140,000.00INR / Each
COMPARE
ASTM 85H Precision, Individually Calibrated 0.800/0.850 Specific Gravity Hydrometer
₹149,000.00INR / Each
COMPARE
ASTM 86H Precision, Individually Calibrated 0.850/0.900 Specific Gravity Hydrometer
₹136,000.00INR / Each
COMPARE
ASTM 89H Precision, Individually Calibrated 1.000/1.050 Specific Gravity Hydrometer
₹160,000.00INR / Each
COMPARE
ASTM 90H Precision, Individually Calibrated 1.050/1.100 Specific Gravity Hydrometer
₹160,000.00INR / Each
COMPARE
ASTM 111H Precision, Individually Calibrated 1.000/1.050 Specific Gravity Hydrometer for Heavy Liquids
₹149,000.00INR / Each
COMPARE
ASTM 113H Precision, Individually Calibrated 1.100/1.150 Specific Gravity Hydrometer for Heavy Liquids
₹136,000.00INR / Each
COMPARE
ASTM 114H Precision, Individually Calibrated 1.150/1.200 Specific Gravity Hydrometer for Heavy Liquids
₹136,000.00INR / Each
COMPARE
ASTM 115H Precision, Individually Calibrated 1.200/1.250 Specific Gravity Hydrometer for Heavy Liquids
₹149,000.00INR / Each
Showing 1- 10 of 10