Cole-Parmer® Essentials Bottletop Dispensers, Recirculation Valve
Save money by conserving reagents
  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows you to change the dispensing angle
These bottletop dispensers conserve reagents with a built-in recirculation valve allowing you to prime and purge without waste. The PTFE piston with ETP o-ring provide frictionless movement reducing the occurrence of bubbles in your dispenses. The springless PTFE valve manifold offers superior chemical compatibility and resists jamming.
55,900.00 - ₹89,300.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Volume (mL) Reproducibility
Availability Pricing
COMPARE
±0.6% 2.5 ±0.2%
₹55,900.00INR / Each
COMPARE
±0.6% 5 ±0.2%
₹65,800.00INR / Each
COMPARE
±0.6% 10 ±0.2%
₹55,900.00INR / Each
COMPARE
±0.6% 30 ±0.2%
₹87,400.00INR / Each
COMPARE
±0.6% 60 ±0.2%
₹87,400.00INR / Each
COMPARE
±0.6% 100 ±0.2%
₹89,300.00INR / Each