Cole-Parmer® Autoclavable Fermentation pH Electrodes
Double-junction electrodes
  • Sealed, gel-filled, double-junction reference electrodes require no refilling
  • Offers long-term stability
  • Extends service life
These electrodes are designed for insertion in RADIOMETER mountings or Ingold nonpressurized assemblies 761-351, 761-31, 761-361, and 361CH-31 (12 mm diameter). Annular ceramic junction eliminates clogging problems and provides excellent stability. Operating temperature range is 23 to 266°>F (-5 to 130°>C). Order detachable cable separately to autoclave your glass-bodied electrode and vessel as a single unit -- simply detach the cable and place a protective cap over the cable connector before sterilizing.
46,300.00 - ₹52,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Autoclavable pH Probe, Refillable/DJ/Glass; 150 mm
₹46,900.00INR / Each
COMPARE
Autoclavable pH Probe, Refillable/DJ/Glass; 210 mm
₹46,300.00INR / Each
COMPARE
Autoclavable pH Probe, Refillable/DJ/Glass; 250 mm
₹47,100.00INR / Each
COMPARE
Autoclavable pH Probe, Refillable/DJ/Glass; 420 mm
₹52,400.00INR / Each