Cole-Parmer Autoclavable Ergonomic Pipettors
For a limited time, while supplies last!
Ergonomic, lightweight design and soft plunger movement reduce hand fatigue
  • Fully autoclavable — no disassembly required
  • Improved accuracy, precision, and performance
These fully autoclavable pipettors are equipped with a stainless steel piston for improved accuracy, precision, and performance. Two-stage plunger operation provided for last-drop blow-out and reverse pipeting. Separate, streamlined tip ejector designed for access into narrow-neck containers and tubes.
Individually calibrated in accordance with DIN 12650 Standards and supplied with a computer-generated calibration report.
9,500.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy (%) Max Volume (µL) Description
Availability Pricing
COMPARE
±1.30% to ±0.50% 1000 Autoclavable Adjustable-Volume Pipette, 100-1000 uL; Each
₹9,500.00INR / Each