This product is now discontinued, View Related Products
Cole-Parmer Analog Stirring Hot Plates
Intuitive controls save you time
  • Compact footprint won't take up valuable benchtop or storage space
  • Warning indicator lights up when top plate is hot, and overheat protection cuts power in case of fault
  • Powerful magnets provide strong stirring action
These general-purpose analog stirring hot plates are designed for performance and safety. The space-saving design takes up less space on the bench or in storage. Powerful magnets and motor support speeds up to 2000 rpm and large volume mixing. All models include an integral fitting for attaching a support rod.
Stirring hot plates (04805-26 & -28, 04807-12 & -14) have either coated aluminum for quick heat-up times and optimal temperature uniformity or cool-edged glass ceramic that is chemically resistant. Hot top plate warning light will flash when the temperature is above 50°C, even when the hot plate power is off (up to 30 minutes duration without power).
246,000.00 - ₹335,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Material
Availability Pricing
COMPARE
120 Aluminum
₹260,000.00INR / Each
COMPARE
230 Aluminum
₹327,000.00INR / Each
COMPARE
120 Ceramic
₹246,000.00INR / Each
COMPARE
230 Ceramic
₹335,000.00INR / Each