Cole-Parmer® CH-400 Series Air-Cooled Portable Industrial Chillers
Chill lasers, electron microscopes and other instruments demanding fluid cooling
  • High capacity in a robust-yet-compact design
  • Chilling-specific, intelligent controller provides fault condition warnings
  • Easy to use and maintain
Choose a sleek portable chiller engineered to accommodate laboratory and industrial process fluid cooling applications. Quickly access the reservoir, filters and all internal components. Temperature is displayed in conjunction with pressure or flow rate on easily visible LED; adjustable alarms enhance user monitoring. Ozone-friendly and air-cooled.
2,706,000.00 - ₹2,869,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (L/min) Power (Hz) Power (VAC)
Availability Pricing
COMPARE
13.2 60 230
₹2,706,000.00INR / Each
COMPARE
41.6 60 230
₹2,869,000.00INR / Each