This product is now discontinued, View Related Products
Cole-Parmer Advanced Digital Stirring Hot Plates
Simple and accurate control
  • Digital display shows surface temperature
  • Warning indicator lights up when top plate is hot (above 50°C) and stays lit when power is off
  • Compact space-saving design won't clutter your workspace
This stirring hot plate offers the convenience of safe storage stacked on its side — great for academic labs. Optional digital temperature controller (order separately) controls the heating of your sample, increasing the accuracy and repeatability of your sample process. Integral fitting accommodates a support rod.
108,000.00 - ₹181,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Material
Availability Pricing
COMPARE
120 Glass Ceramic
₹181,000.00INR / Each
COMPARE
230 Glass Ceramic
₹179,000.00INR / Each
COMPARE
120 Coated Aluminum/Silicon
₹108,000.00INR / Each
COMPARE
230 Coated Aluminum/Silicon
₹161,000.00INR / Each