This product is now discontinued, View Related Products
Cole-Parmer Advanced Analog Stirring Hot Plates
Intuitive controls save you time
  • Compact footprint won't take up valuable benchtop or storage space
  • Accurate temperature control with colored LED indicator scale
  • Warning indicator lights up when top plate is hot, and overheat protection cuts power in case of fault
  • Powerful magnets provide strong stirring action
These general-purpose analog stirring hot plates are designed for performance and safety. The space-saving design takes up less space on the bench or in storage. Powerful magnets and motor support speeds up to 2000 rpm and large volume mixing. All models include an integral fitting for attaching a support rod.
Advanced stirring hot plates (04801-60 to -67) feature either coated aluminum for quick heat-up times and optimal temperature uniformity or cool-edged glass ceramic that is chemically resistant. Hot top plate warning light will flash when the temperature is above 50°C, even when the hot plate power is off (up to 30 minutes duration without power). An innovative LED temperature indicator scale lights up in colored increments as the top plate heats up, and shows you when the plate has reached its temperature set point. Order the digital temperature controller (04801-80) to accurately regulate sample temperature.
126,000.00 - ₹177,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Material
Availability Pricing
COMPARE
120 Glass Ceramic
₹177,000.00INR / Each
COMPARE
230 Glass Ceramic
₹173,000.00INR / Each
COMPARE
120 Coated Aluminum/Silicon
₹126,000.00INR / Each
COMPARE
230 Coated Aluminum/Silicon
₹155,000.00INR / Each