This product is now discontinued, View Related Products
Cole-Parmer® 125 Tissue Homogenizer Kits
Handle small tissue samples, PCR, RNA, and DNA up to heavy-duty tissue processing and cell disruption
  • Variable-speed motor performs up to 30,000 rpm
  • Homogenize tissue samples quickly and efficiently — without worrying about cross-contamination
Cole-Parmer homogenizers are well-suited for a wide range of processing applications —use as a handheld unit or post-mount to the optional stand for hands-free operation. Plastic tip probes are maintenance-free and do not require the disassembly associated with traditional stainless steel generators. Tips are available in both soft tissue (deaggregations) and hard tissue (frozen) versions; designed to work with sample volumes between 0.25 to 30 mL. Engineered from durable plastics, these tips may be autoclaved and reused, or disposed of after each use to eliminate cross contamination.
197,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Min Sample Size (mL) Power (VAC) Description
Availability Pricing
COMPARE
0.25 115 125 Homogenizer Kit, Hard tissue version
₹197,000.00INR / Each
COMPARE
0.25 220 125 Homogenizer Kit, Hard tissue version, 220 VAC
₹197,000.00INR / Each
COMPARE
0.25 115 125 Homogenizer Kit, Soft tissue version, 115 VAC
₹197,000.00INR / Each
COMPARE
0.25 220 125 Homogenizer Kit, Soft tissue version, 220 VAC
₹197,000.00INR / Each