Cole-Parmer 12-mm pH Electrodes with Integrated Assebblies
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersible
Great for routine pH applications. Use blue glass bulb for measurements up to 13 pH. Electrodes withstand temperatures from 23 to 212ºF (-5 to 100ºC).
21,900.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Removable General-Purpose, Single-Junction pH Electrode
₹21,900.00INR / Each