Erie Scientific Clipped-Corner Slides
Corners are clipped for easy handing
  • Clipped corners keep sample away from the edges of the slide—ideal for making smears
Drastically reduce the chances of cutting your finger or tearing a glove when you use these slides. Slides measure 75 x 25 mm and are 1-mm thick.
7,500.00 - ₹11,400.00INR / Pkg of 144
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size Slide Type Description
Availability Pricing
Erie Scientific Clipped-Corner Slides, Plain Glass, 144/Pk WW-48520-01 Mfr # 2950-4-001
COMPARE
75 x 25 mm Plain Clipped-Corner Slides, plain glass, 144/PK
₹7,500.00INR / Pkg of 144
Erie Scientific Clipped-Corner Slides, Superfrost, 144/Pk WW-48520-05 Mfr # 4951-4-001
COMPARE
75 x 25 mm Superfrost Clipped-Corner Slides, Superfrost, 144/PK
₹11,400.00INR / Pkg of 144