Thermo Scientific™ Sample Glass Vial Kits
Save time during sample preparation and reduce the risk of contamination
  • Choose from clear or amber glass vials
Screw-thread sample vials are manufactured from either clear 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass. Vials maintain PH level and eliminate ion leaching. Choose from a polypropylene cap with a PTFE-lined foam backing or a polypropylene cap with a PTFE-lined silicone septa. Vials, caps, and septa are shrink-wrapped separately.
12,200.00 - ₹52,000.00INR / Pkg of 200
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Material Tube Material Volume (mL)
Availability Pricing
Thermo Scientific B7800-2 Vial Kit, Clear Glass, Polypropylene Cap with PTFE Liner, 4 mL; 100/Pk WW-08922-22 Mfr # 6AK4CCT
COMPARE
Urethane Borosilicate Glass 4
₹12,200.00INR / Pkg of 100
Thermo Scientific B7800-12 Vial Kit, Clear Glass, Polypropylene Cap with PTFE Liner, 12 mL; 200/Pk WW-08922-24 Mfr # 6AK12CCT
COMPARE
Urethane Borosilicate Glass 12
₹32,500.00INR / Pkg of 200
Thermo Scientific B7800-4 Vial Kit, Clear Glass, Polypropylene Cap with PTFE Liner, 16 mL; 200/Pk WW-08922-26 Mfr # 6AK16CCT
COMPARE
Polypropylene Borosilicate Glass 16
₹34,800.00INR / Pkg of 200
Thermo Scientific B7800-2A Vial Kit, Amber Glass, Polypropylene Cap with PTFE Liner, 4 mL; 100/Pk WW-08922-32 Mfr # 6AK4ACT
COMPARE
Urethane Borosilicate Glass 4
₹23,000.00INR / Pkg of 100
Thermo Scientific B7800-3A Vial Kit, Amber Glass, Polypropylene Cap with PTFE Liner, 8 mL; 200/Pk WW-08922-33 Mfr # 6AK8ACT
COMPARE
Urethane Borosilicate Glass 8
₹29,400.00INR / Pkg of 200
Thermo Scientific B7800-12A Vial Kit, Amber Glass, Polypropylene Cap with PTFE Liner, 12 mL; 200/Pk WW-08922-34 Mfr # 6AK12ACT
COMPARE
Urethane Borosilicate Glass 12
₹33,500.00INR / Pkg of 200
Thermo Scientific B7800-6A Vial Kit, Amber Glass, Polypropylene Cap with PTFE Liner, 40 mL; 100/Pk WW-08922-39 Mfr # 6AK40ACT
COMPARE
Urethane Borosilicate Glass 40
₹23,800.00INR / Pkg of 100
Thermo Scientific B7990-2 Vial Kit, Clear Glass, Polypropylene Cap with PTFE/Silicone Septa, 4mL; 100/Pk WW-08922-42 Mfr # B7990-2
COMPARE
Polypropylene Borosilicate Glass 4
₹13,900.00INR / Pkg of 100
Thermo Scientific B7990-3 Vial Kit, Clear Glass, Polypropylene Cap with PTFE/Silicone Septa, 8 mL; 200/Pk WW-08922-43 Mfr # B7990-3
COMPARE
Polypropylene Borosilicate Glass 8
₹32,500.00INR / Pkg of 200
Thermo Scientific B7990-12 Vial Kit, Clear Glass, Polypropylene Cap with PTFE/Silicone Septa, 12 mL; 200/Pk WW-08922-44 Mfr # B7990-12
COMPARE
Polypropylene Borosilicate Glass 12
₹32,300.00INR / Pkg of 200
Thermo Scientific B7990-4 Vial Kit, Clear Glass, Polypropylene Cap with PTFE/Silicone Septa, 16 mL; 200/Pk WW-08922-46 Mfr # B7990-4
COMPARE
Polypropylene Borosilicate Glass 16
₹42,900.00INR / Pkg of 200
Thermo Scientific B7990-5 Vial Kit, Clear Glass, Polypropylene Cap with PTFE/Silicone Septa, 22 mL; 200/Pk WW-08922-48 Mfr # 6AK20COTAS
COMPARE
Polypropylene Borosilicate Glass 22
₹52,000.00INR / Pkg of 200
Thermo Scientific B7990-3A Vial Kit, Amber Glass, Polypropylene Cap with PTFE/Silicone Septa, 8 mL; 200/Pk WW-08922-53 Mfr # B7990-3A
COMPARE
Polypropylene Borosilicate Glass 8
₹35,500.00INR / Pkg of 200
Thermo Scientific B7990-12A Vial Kit, Amber Glass, Polypropylene Cap with PTFE/Silicone Septa, 12 mL; 200/Pk WW-08922-54 Mfr # B7990-12A
COMPARE
Polypropylene Borosilicate Glass 12
₹37,200.00INR / Pkg of 200
Showing 1- 14 of 14