This product is now discontinued, View Related Products
Corning Chromatography Columns with Reservoirs
Ideal for adsorption, gel, permeation, and ion exchange techniques
  • Perfect for flash chromatography applications
  • Easily control the flow rate
LabGlass Chromatography Columns with attached reservoirs are perfect for flash chromatography applications. Columns include a PTFE plug to control flowrate. Standard joint size is TS 24/40. Also available with coarse fritted TS 35/25 joint size; call our Application Specialists for details.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Description
Availability Pricing
Corning Glass Chromatography Column with Reservoir, 200 mL WW-34695-54 Mfr # LG4564T-100
COMPARE
200 Glass Chromatography Column with Reservoir, 200 mL
Call For Price
Corning Glass Chromatography Column with Reservoir, 250 mL WW-34695-56 Mfr # LG4564T-104
COMPARE
250 Glass Chromatography Column with Reservoir, 250 mL