This product is now discontinued, View Related Products
Chemware Narrow-Mouth Bottles, PFA
Non-contaminating, lowest metal ion content of all fluropolymers
Durable leakproof PFA bottles are ideal for environmental sampling and storage in pharmaceutical and semiconductor processing. Chemically inert and heat resistant bottles store almost all corrosive chemicals. Includes caps with fluoropolymer cap liner.
211,000.00INR / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Neck Inside Diameter (ID) (mm)
Availability Pricing
Chemware D1069068 Narrow-Mouth Graduated PFA Bottle, 100 mL (3 oz), 1/Pk WW-06040-30 Mfr # D1069068
COMPARE
100 50 15.9
Call For Price
Chemware D1069071 Narrow-Mouth Graduated PFA Bottle, 250 mL (8 oz), 1/Pk WW-06040-32 Mfr # D1069071
COMPARE
250 50 15.9
Call For Price
Chemware D1069072 Narrow-Mouth Graduated PFA Bottle, 500 mL (16 oz), 1/Pk WW-06040-34 Mfr # D1069072
COMPARE
500 100 17
Call For Price
Chemware D1069069 Narrow-Mouth Graduated PFA Bottle, 1 L (32 oz), 1/Pk WW-06040-36 Mfr # D1069069
COMPARE
1000 100 25.4
Call For Price
Chemware D1069086 Narrow-Mouth Graduated PFA Bottle, 2 L (68 oz), 1/Pk WW-06040-38 Mfr # D1069086
COMPARE
2000 100
Call For Price
Chemware D1069073 Narrow-Mouth Graduated PFA Bottle, 3 L (96 oz), 1/Pk WW-06040-40 Mfr # D1069073
COMPARE
3000 500 36.6
₹211,000.00INR / Pkg of 1
Chemware D1069074 Narrow-Mouth Graduated PFA Bottle, 5 L (160 oz), 1/Pk WW-06040-42 Mfr # D1069074
COMPARE
5000 1000 36.4
Call For Price
Chemware D1069787 Narrow-Mouth Graduated PFA Bottle, 10 L (320 oz), 1/Pk WW-06040-44 Mfr # D1069787
COMPARE
10000 0 36.4
Call For Price
Showing 1- 8 of 8