Chemical Resistant PFA Trays
These compact-size, chemical resistant trays are perfect to use for corrosive or solvent baths. Trays can withstand temperatures up to 500°>F (260°>C).
Use model 06715-10 for staining 25 mm x 75 mm slides and holding other small items. The included cover prevents evaporation. Use model 06715-20 as a sterilization tray or to hold containers with corrosive contents to protect your bench from spills. Cover not included. CHEMWARE®.
39,800.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (in)
Availability Pricing
Chemware D1069686 PFA Tray without Cover, 6 x 4 x 2" WW-06715-20 Mfr # D1069686
COMPARE
6
₹39,800.00INR / Each