Chem-Tainer UN Spec (1H1) Tight-Head Seamless Drums
No more heavy steel drums
  • Withstand rough handling—lightweight, rugged design
  • Easy access through 2" and 3⁄4" NPT bungs
  • Tough sides can support mixer
Tight-head drums are 1H1 single packages complying with UN specifications and meet HM-181 and IATA Specification Packaging Standards. These drums are constructed of durable LLDPE for a wide range of applications.
53,500.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Volume (Gal) Wall thickness (in)
Availability Pricing
Chem-Tainer SH192930 UN Specification (1H1) Tight-Head Drum, 30 Gal. WW-06950-40 Mfr # SH192930
COMPARE
LLDPE 30 0.19
₹53,500.00INR / Each