CELLTREAT® Bacteriological/Milk Pipettes
Meet APHA requirements for Standard Methods and ASTM E1043-00(2010)
  • Comply with Standards Methods for Examination of Dairy Products and ASTM Standard Specification for Pipets, Milk and Cream Examination
  • Choose from individually or bulk wrapped, in 1.1- or 2.2-mL sizes
  • All pipettes have been gamma irradiated for sterilization
  • Clear polystyrene construction is durable, disposable, and easily seen through for accurate readings
42,700.00 - ₹45,500.00INR / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Package Of
Availability Pricing
CELLTREAT Scientific Products 229257 Bacteriological/Milk Pipette, 1.1 mL, Bulk Packed, Sterile; 500/Cs WW-34567-72 Mfr # 229257
COMPARE
1.1 50 bulk packed per bag
₹42,700.00INR / Case of 500
CELLTREAT Scientific Products 229258 Bacteriological/Milk Pipette, 2.2mL, Bulk Packed, Sterile; 400/Cs WW-34567-73 Mfr # 229258
COMPARE
2.2 50 bulk packed per bag
₹45,500.00INR / Case of 400