Cole-Parmer® Essentials Cell and Tissue Culture Dishes
Excellent optical quality helps to promote healthy cells and reproducible results
  • Select from non-treated and surface-treated dishes with 7 different diameters of 1.8, 3.5, 6.0, 7.0, 9.0, 10.0, and 15.0 cm
  • Flat bottom with uniform wall thickness ensures stability
  • Corrugation free and smooth dish surface maximizes usable growth area
  • The rim on upper side of the lid mates with the dish brim for easy and secure stacking
  • Lids with vents are available for very effective gas exchange
  • Gamma sterilized and non-pyrogenic
  • Packaged in durable zip re-sealable self-standing plastic bags that allow dishes to remain upright and reduce contamination
  • Inner packaging is individually labelled for lot traceability
10,300.00 - ₹27,500.00INR / Case of 1
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
10 dishes/bag; 96 bags/case Total: 960 dishes
₹23,200.00INR / Case of 1
COMPARE
10 dishes/bag; 60 bags/case Total: 600 dishes
₹16,300.00INR / Case of 1
COMPARE
10 dishes/bag; 60 bags/case Total: 600 dishes
₹16,300.00INR / Case of 1
COMPARE
10 dishes/bag; 50 bags/case Total: 500 dishes
₹17,200.00INR / Case of 1
COMPARE
10 dishes/bag; 30 bags/case Total: 300 dishes
₹10,300.00INR / Case of 1
COMPARE
5 dishes/bag; 20 bags/case Total: 100 dishes
₹14,000.00INR / Case of 1
COMPARE
1 dish/bag; 120 bags/case Total: 120 dishes
₹19,600.00INR / Case of 1
COMPARE
10 dishes/bag; 96 bags/case Total: 960 dishes
₹27,500.00INR / Case of 1
COMPARE
10 dishes/bag; 60 bags/case Total: 600 dishes
₹21,400.00INR / Case of 1
COMPARE
10 dishes/bag; 60 bags/case Total: 600 dishes
₹20,200.00INR / Case of 1
COMPARE
10 dishes/bag; 50 bags/case Total: 500 dishes
₹24,000.00INR / Case of 1
COMPARE
10 dishes/bag; 30 bags/case Total: 300 dishes
₹14,400.00INR / Case of 1
COMPARE
5 dishes/bag; 20 bags/case Total: 100 dishes
₹17,300.00INR / Case of 1
COMPARE
1 dish/bag; 120 bags/case Total: 120 dishes
₹23,500.00INR / Case of 1
Showing 1- 14 of 14