CDS Analytical Empore™ SPE 96-Well Plates

Offers efficient way to purify bio-analytical samples
  • Examine multiple parameters within a single plate
  • Reduces time required for method optimization
  • Compatible with standard microtiter liquid handling technologies and injection systems
  • Fits standard 96-well plate format
The Empore plates come with 96 wells, ideal for solid phase extraction (SPE) of analytes. The well plates provide an efficient method to purify a variety of bio-analytical samples and offer robust analytical method development. Parallel sample processing allows 96 samples to be extracted within one hour. Each well contains a particle-loaded membrane for efficient sample extraction followed by elution in as little as 100 L with mobile phase or mobile phase compatible solutions. The small bed mass of sorbent in the well plates allow you to use smaller solvent volumes than with traditional packed bed SPE products. Sorbent phases include C8, C18, MPC (Mixed Phase Cation). The plates are compatible with vacuum manifold and well plate covers. Use sealing tape pads to cover unused well during an extraction assay.
253,000.00 - ₹975,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Well Volume (mL) Description
Availability Pricing
CDS Analytical  6015SD Empore™ 96-Well Plate, C18 Silica, 50um, 1.2mL WW-35213-06 Mfr # G2396SD
COMPARE
96 1.2 96-well Plate, C18 Silica, 50um, 1.2mL
₹975,000.00INR / Each
CDS Analytical  6014SD Empore™ 96-Well Plate, C8 Silica, 50um, 1.2mL WW-35213-07 Mfr # G2296SD
COMPARE
96 1.2 96-well Plate, C8 Silica, 50um, 1.2mL
₹975,000.00INR / Each
CDS Analytical  6030SD Empore™ 96-Well Plate, MPC, 50um, 1.2mL WW-35213-08 Mfr # G2096SD
COMPARE
96 1.2 96-well Plate, MPC, 50um, 1.2mL
₹975,000.00INR / Each
CDS Analytical  6065 Empore™ 96-Well Plate, PP Filter, 5.5mm dia., 1.2mL WW-35213-37 Mfr # G2800
COMPARE
96 1.2 96-well Plate, PP, 5.5mm dia., 1.2mL
₹253,000.00INR / Each
CDS Analytical  6360 Empore™ 96-Well Plate, PP Filter, 5.5mm dia., 2.5mL WW-35213-38 Mfr # G2800DW
COMPARE
96 2.5 96-well Plate, PP, 5.5mm dia., 2.5mL
₹261,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat