CDS Analytical Empore™ RAD SPE Disks
Efficient alternative to conven¬tional radiochemical sample preparation methods
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
The Empore RAD disks provide a large surface area for sorbent/sample contact, idea for use with purification and concentration of analytes from large volume aqueous samples. A proprietary process is used to entrap adsorbent particles into a matrix of inert PTFE to create a mechanically stable sorbent disk.
In addition to fast flow rates and high throughput, the disks form convenient direct counting sources when used in applicable test methods. Disks are available in four selective sorbents – cesium, radium, strontium, and technetium.
416,000.00 - ₹2,214,000.00INR / Pkg of 60
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Description
Availability Pricing
CDS Analytical  3291 Empore™ Radium SPE RAD Disk, 47mm; 60/PK WW-35213-11 Mfr # G2647
COMPARE
47 Radium Empore™ Radium Rad Disk, 47mm; 60/PK
₹2,214,000.00INR / Pkg of 60
CDS Analytical  3292 Empore™ Technetium SPE RAD Disk, 47mm; 60/PK WW-35213-12 Mfr # G2747
COMPARE
47 Technetium Empore™ Technetium Rad Disk, 47mm; 60/PK
₹416,000.00INR / Pkg of 60
CDS Analytical  3290 Empore™ Strontium SPE RAD Disk, 47mm; 60/PK WW-35213-13 Mfr # G2847
COMPARE
47 Strontium Empore™ Strontium Rad Disk, 47mm; 60/PK
₹2,056,000.00INR / Pkg of 60
CDS Analytical  3293 Empore™ Cesium SPE RAD Disk, 47mm; 60/PK WW-35213-14 Mfr # G2947
COMPARE
47 Cesium Empore™ Cesium Rad Disk, 47mm; 60/PK
₹1,070,000.00INR / Pkg of 60