This product is now discontinued, View Related Products
CDI Quick-Set Split-Beam Torque Wrenches
Rugged design is ideal for any environment
  • Split-beam measuring element provides accurate, reliable readings and eliminates heavy coil spring used in conventional click wrenches
  • Accurate within ±4% of any clockwise setting from 20% of full scale to full scale
  • Sealed head design flexes up to 15°, providing knuckle and obstruction clearance
  • Thumb screw adjustment allows rapid cycling through micrometer-style wrenches
  • Guard prevents accidental setting changes
  • Conversion table to N-m is displayed on the handle
  • Sealed neck keeps dust and grit away from torque mechanism for longer tool life
  • Fewer moving parts reduces friction and wear
  • Ergonomic cushion grip handle provides comfort and control and resists most fluids
These unique split-beam torque wrenches provide consistent, accurate readings in a sturdy maintenance-free design. Wrenches allow you to quickly adjust settings by rotating the knob until the required value is displayed in the torque window. The unique sealed ratchet head requires almost no maintenance — allowing you to spend more time working and less time on maintenance. Wrenches are designed for measuring torque in a clockwise direction only (not reversible).
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-ft)
Availability Pricing
J. H. Williams Tool Group WSC-100CA Torque Wrench, Split Beam, 20 to 100 Lbs. WW-25301-26 Mfr # WSC-100CA
COMPARE
100
Call For Price
J. H. Williams Tool Group WSC-250CA Torque Wrench, Split Beam, 45 to 250 Lbs. WW-25301-27 Mfr # WSC-250CA
COMPARE
250
Call For Price
J. H. Williams Tool Group WSC-600CA Torque Wrench, Split Beam, 200 to 600 Lbs. WW-25301-28 Mfr # WSC-600CA
COMPARE
600