Casella dBadge Micro Noise Dosimeters

Easily position on clothing or hard hats
  • Lightweight less than 2.5 oz
  • Small size—only 2.8" long, 1.7" wide and 2" tall
  • Quick charge in less than 1.5 hours from completely empty
  • Store all results for later review
  • Calculate all the popular noise dose indices
  • Time history profiling of 4 results at fixed 1 min intervals
  • Measure 3 dB and 5 dB exchange rates simultaneously
  • Lockable in operation mode
  • Direct automatic calibration with standard acoustic
  • calibrator (1⁄2" cavity)
  • Intrinsically safe model (98933-35) allows use in hazardous areas—these units have worldwide ATEX approval
The CEL350 has all the advantages of earlier badge designs, such as small size and light weight, and yet it still provides all of the benefits that users have come to expect from a personal noise dosimeter. The CEL350 is fully
featured and able to satisfy all current noise measurement protocols during a single run—an ideal instrument for both experienced and new users of dosimeters. Unit stores all results for later investigation and reporting. Easy, push-button setup.
493,000.00INR / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Casella CEL-350IS Individual Intrinsically Safe micro noise badge, with mounting clips WW-98933-35 Mfr # dBadge2IS
COMPARE
Micro Noise Badge Intrinsically Safe with Mounting Clips and Windscreenr Only
₹493,000.00INR / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat