This product is now discontinued, View Related Products

Casella dBadge Micro Noise Dosimeter Kits

More convenient than the traditional noise dosimeter with remote cable microphone
  • Lightweight less than 2.5 oz
  • Small size—only 2.8" long, 1.7" wide and 2" tall
  • Quick charge in less than 1.5 hours from completely empty
  • Stores all results for later review
  • Calculates all the popular noise dose indices
  • Time history profiling of 4 results at fixed 1 min intervals
  • Wide dynamic range from 65 to a full 140 dB
  • Measures simultaneously with 3 dB and 5 dB exchange rates
  • Lockable in operation mode
  • Direct automatic calibration with standard acoustic calibrator (1/2" cavity)
  • Intrinsically safe model allows use in hazardous area such as petrochemical plants. This unit has worldwide ATEX approval
The CEL350 has all the advantages of earlier badge designs, such as small size and light weight, and yet it still provides all of the benefits that users have come to expect from a personal noise dosimeter. The CEL350 is fully featured and able to satisfy all current noise measurement protocols during a single run—an ideal instrument for both experienced and new users of dosimeters. Unit stores all results for later investigation and reporting. Easy, push-button setup.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Decibel (Db) High Range (dB) Function Description
Availability Pricing
Casella CEL-350/K10 Micro Noise Dosimeter Kit , 10 Pk WW-50509-38 Mfr # dBadge2/K10
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Micro Noise Dosimeter Kit , 10 Pk
Call For Price
Casella CEL-350-K1 Micro Noise Dosimeter Kit WW-50509-39 Mfr # dBadge2/K1
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Micro Noise Dosimeter Kit
Call For Price
Casella CEL-350/K2 Micro Noise Dosimeter Kit , 2Pk WW-86536-01 Mfr # dBadge2/k2
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Micro Noise Dosimeter Kit , 2Pk
Call For Price
Casella CEL-350/K5 Micro Noise Dosimeter Kit , 5Pk WW-86536-03 Mfr # dBadge2/k5
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Micro Noise Dosimeter Kit , 5Pk
Call For Price
Casella CEL-350IS-K1 Noise Dosimeter Kit intrinsically Safe WW-86536-13 Mfr # dBadge2IS/K1
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Noise Dosimeter Kit intrinsically Safe
Call For Price
Casella CEL-350IS-K2 Noise Dosimeter Kit intrinsically Safe, 2Pk WW-86536-14 Mfr # dBadge2IS/K2
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Noise Dosimeter Kit intrinsically Safe, 2Pk
Call For Price
Casella CEL-350IS-K5 Micro Noise Dosimeter Kit intrinsically Safe, 5Pk WW-86536-15 Mfr # dBadge2IS/K5
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Micro Noise Dosimeter Kit intrinsically Safe, 5Pk
Call For Price
Casella CEL-350IS-K10 Micro Noise Dosimeter Kit intrinsically Safe, 10Pk WW-86536-16 Mfr # dBadge2IS/K10
COMPARE
140 Lavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose% Micro Noise Dosimeter Kit intrinsically Safe, 10Pk
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat