Carbon-Impregnated Pleated Polyester Cartridges for "Big Blue" Housing
The pleated design offers higher flow rates and dirt-loading capacities than shorter-life block cartridges. The non-cellulose media resists chemical attack and biofilm build-up making the filter ideal for both chlorinated and non-chlorinated water treatment applications. Common applications include post reverse osmosis polishing and well water treatment.
6,300.00 - ₹11,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Micron Rating
Availability Pricing
Big Blue Filter cartridge, 10", pleated carbon, 10 micron WW-29830-32 Mfr # 155398-43(NCP-BB)
COMPARE
10 10
₹6,300.00INR / Each
Big Blue Filter cartridge, 20", pleated carbon, 10 micron WW-29830-33 Mfr # 155382-43(NCP-20BB)
COMPARE
20 10
₹11,700.00INR / Each