Carbon Fiber Digital Fractional Caliper
Versatility and accuracy in a compact design
  • Lightweight, non-marring construction
  • Tri-Mode digital display – inches, millimeters, and fractions
  • For making inside, outside, depth and step measurements
  • Auto on/off feature preserves battery life
  • Compact, pocket-sized design
  • Bar scale is graduated in inches and millimeters
  • Free-wheeling friction roller advances and retracts slide
  • Extendable depth gauge for use in slots and small holes
These carbon fiber digital calipers measure inside diameter, outside diameter, depth, and step measurements. Easily switch from inches to millimeters to fractions with the touch of a button. Fine-adjustment thumb roller improves reading speed and position-holding lock screw sets length.
4,400.00 - ₹6,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in) Description
Availability Pricing
General Tools 1433 Light Weight Carbon Fiber 0-3" digital calipers WW-97152-01 Mfr # 1433
COMPARE
3 Light Weight Carbon Fiber 0-3\" digital calipers
₹4,400.00INR / Each
COMPARE
6 Lightweight Carbon Fiber 0-6\" Digital Caliper
₹6,600.00INR / Each