Capsette Cassettes
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Capsette cassettes accept standard stainless steel lids
9,500.00INR / Pkg of 500
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Color
Availability Pricing
Bio Plas Capsette 6030 Cassettes, White, 500/pack WW-48620-00 Mfr # 6030
COMPARE
White
₹9,500.00INR / Pkg of 500