Cannon®-Fenske Calibrated Glass Capillary Opaque Viscometers
Determine viscosities using ASTM testing methods
  • Each viscometer is calibrated in accordance with ASTM D446 Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
  • Reverse flow type
Use for dark Newtonian liquids; especially suited for liquids so dark in color that the meniscus cannot be seen in a Cannon-Fenske routine viscometer. Use to study the effects of various additives on lubricating and hydraulic oils. Also use to study shearing stress and shearing rate. Require a 12-mL sample volume.
53,100.00 - ₹59,900.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Approximate Constant (cSt/sec) Max Viscosity Range (cSt) Universal Size Number
Availability Pricing
Cannon 9721-F50 Viscometer, universal size 25 WW-98934-70 Mfr # 9721-F50
COMPARE
0.002 2 25
₹59,900.00INR / Each
Cannon 9721-F53 Viscometer, universal size 50 WW-98934-71 Mfr # 9721-F53
COMPARE
0.004 4 50
₹53,100.00INR / Each
Cannon 9721-F56 Viscometer, universal size 75 WW-98934-72 Mfr # 9721-F56
COMPARE
0.008 8 75
₹53,100.00INR / Each
Cannon 9721-F59 Viscometer, universal size 100 WW-98934-73 Mfr # 9721-F59
COMPARE
0.015 15 100
₹53,100.00INR / Each
Cannon 9721-F62 Viscometer, universal size 150 WW-98934-74 Mfr # 9721-F62
COMPARE
0.035 35 150
₹53,100.00INR / Each
Cannon 9721-F65 Viscometer, universal size 200 WW-98934-75 Mfr # 9721-F65
COMPARE
0.1 100 200
₹53,100.00INR / Each
Cannon 9721-F68 Viscometer, universal size 300 WW-98934-76 Mfr # 9721-F68
COMPARE
0.25 250 300
₹53,100.00INR / Each
Cannon 9721-F71 Viscometer, universal size 350 WW-98934-77 Mfr # 9721-F71
COMPARE
0.5 500 350
₹53,100.00INR / Each
Cannon 9721-F74 Viscometer, universal size 400 WW-98934-78 Mfr # 9721-F74
COMPARE
1.2 1200 400
₹53,100.00INR / Each
Cannon 9721-F77 Viscometer, universal size 450 WW-98934-79 Mfr # 9721-F77
COMPARE
2.5 2500 450
₹53,100.00INR / Each
Cannon 9721-F80 Viscometer, universal size 500 WW-98934-80 Mfr # 9721-F80
COMPARE
8 8000 500
₹53,100.00INR / Each
Cannon 9721-F83 Viscometer, universal size 600 WW-98934-81 Mfr # 9721-F83
COMPARE
20 20000 600
₹53,100.00INR / Each
Cannon 9721-F86 Viscometer, universal size 650 WW-98934-82 Mfr # 9721-F86
COMPARE
45 45000 650
₹53,100.00INR / Each
Cannon 9721-F89 Viscometer, universal size 700 WW-98934-83 Mfr # 9721-F89
COMPARE
100 100000 700
₹53,100.00INR / Each
Showing 1- 14 of 14