This product is now discontinued, View Related Products
BW Clip Single Gas Detectors
Compact and easy to use — no maintenance required!
Operated by an advanced microcontroller and electrochemical sensor, the BW Clip monitors provide continuous, maintenance-free operation for up to two years from the day of activation. These disposable battery-operated monitors are easy to use — simply clip the meter to your pocket or belt and go!
View important gas-reading information and detector life remaining data on the full-feature LCD display. Monitors feature audible, visible, and vibrating alarms. Each monitor is encased in a shock- and water-resistant ABS enclosure and offers RFI protection to withstand the rigors of industrial and field use.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
BW Technologies BW Clip Single Gas Detector, H2S (Hydrogen Sulfide), 2-Year WW-86383-20 Mfr # BWC2-H
COMPARE
Single Gas Detector, H2S (Hydrogen Sulfide), 2-Year
Call For Price
BW Technologies BW Clip Single Gas Detector, CO (Carbon Monoxide), 2-Year WW-86383-21 Mfr # BWC2-M
COMPARE
Single Gas Detector, CO (Carbon Monoxide), 2-Year
Call For Price
BW Technologies BW Clip Single Gas Detector, SO2 (Sulfur Dioxide), 2-Year WW-86383-22 Mfr # BWC2-S
COMPARE
Single Gas Detector, SO2 (Sulfur Dioxide), 2-Year
Call For Price
BW Technologies BW Clip Single Gas Detector, O2 (Oxygen), 2-Year WW-86383-23 Mfr # BWC2-X
COMPARE
Single Gas Detector, O2 (Oxygen), 2-Year
Call For Price
BW Technologies BW Clip Single Gas Detector, H2S (Hydrogen Sulfide), 3-Year WW-86383-24 Mfr # BWC3-H
COMPARE
Single Gas Detector, H2S (Hydrogen Sulfide), 3-Year
Call For Price
BW Technologies BW Clip Single Gas Detector, CO (Carbon Monoxide), 3-Year WW-86383-25 Mfr # BWC3-M
COMPARE
Single Gas Detector, CO (Carbon Monoxide), 3-Year