Bud Industries NEMA 4X Fiberglass Enclosure Boxes
Fiberglass-reinforced plastic provides outstanding protection against corrosion
  • Watertight, chemical resistant, durable, and impervious to dents
  • Stainless steel hinge, screws, and wall mounting brackets
  • Includes powder-coated 14-gauge steel removable inner panel
  • Light gray finish
  • NF models have a stainless steel screw-type cover; NFL models have a stainless steel latch-type cover
  • Designed to meet NEMA 1, 2, 4, 4X, 12, 13, and IP65 specifications
  • UL listed and tested to UL508-4X requirements, UL File E194432
14,300.00 - ₹21,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries NF 6610 NEMA 4X Fiberglass Box with Screw Cover and Mounting Brackets 9450180 Mfr # NF-6610
COMPARE
4.8 7.75 7.75
₹14,300.00INR / Each
Bud Industries NF 6612 NEMA 4X Fiberglass Box with Screw Cover and Mounting Brackets 9450184 Mfr # NF-6612
COMPARE
6.8 11.78 9.9
₹21,200.00INR / Each