Bud Industries Diecast Aluminum NEMA 4 Enclosures
Aluminum alloy is resistant to environmental and atmospheric conditions
Aluminum alloy #ADC-10 enclosure is designed to IP65 of IEC 529 and NEMA 4 specifications, and is UL508-4 certified. House sensitive electronic assemblies, pneumatic, hydraulic or electrical devices in wet and dirty commercial and industrial environments. Enclosures have internal bosses for mounting PCBs or connecting terminals, grounds, or other electrical and control devices. Cover incorporates provisions for recessed gasket. Gasket is EPDM. In order to maintain NEMA 4X rating, cable glands must be used when cables enter or exit the enclosure.
6,500.00 - ₹9,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries AN-1305 Diecast aluminum enclosure, type 9450106 Mfr # AN-1305-A
COMPARE
2.95 5.83 4.25
₹6,500.00INR / Each
Bud Industries AN-1307 Diecast aluminum enclosure, type 9450110 Mfr # AN-1307-A
COMPARE
2.17 8.75 5.75
₹9,300.00INR / Each