This product is now discontinued, View Related Products
BriskHeat® Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Tape with X2 PID Temperature Controller Bundles
Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
  • Programmable controller features alarm, user selectable °C or °F, and advanced PID control
  • Easy to read measurements with 0.4” high, 4-digit, green LED display
  • Type J thermocouple provides feedback for strict temperature control
  • Enhanced durability with heavy insulated design
  • Constructed of heavy insulated knitted and braided Somax® fiberglass
  • Highly flexible, multi-stranded, dual heating element provides even heating across tape
All-inclusive bundle features a digital PID controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three button control with a programmable temperature range of 0° to 760°C (32° to 1400°F). This advanced controller offers an accuracy with Type-J heating cords of ±0.5°C (±0.9°F).
The heavy insulated heating tapes feature an IP5X, rating making them ideal for rugged applications. Select heating tape length to optimize coverage while avoiding overlap.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Width (in) Description
Availability Pricing
BriskHeat X2J-BWH051020LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 2.0 ft; 120 VAC WW-36120-59 Mfr # X2J-BWH051020LD
COMPARE
2 1/2 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 2.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051040LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 4.0 ft; 120 VAC WW-36120-60 Mfr # X2J-BWH051040LD
COMPARE
4 1/2 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 4.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051060LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 6.0 ft; 120 VAC WW-36120-61 Mfr # X2J-BWH051060LD
COMPARE
6 1/2 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 6.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051080LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 8.0 ft; 120 VAC WW-36120-62 Mfr # X2J-BWH051080LD
COMPARE
8 1/2 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 8.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051100LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 10.0 ft; 120 VAC WW-36120-63 Mfr # X2J-BWH051100LD
COMPARE
10 1/2 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in X 10.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH101020LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in X 2.0 ft; 120 VAC WW-36120-64 Mfr # X2J-BWH101020LD
COMPARE
2 1 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in X 2.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH101040LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in X 4.0 ft; 120 VAC WW-36120-65 Mfr # X2J-BWH101040LD
COMPARE
4 1 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in X 4.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH101060LD Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in X 6.0 ft; 120 VAC WW-36120-66 Mfr # X2J-BWH101060LD
COMPARE
6 1 Heavy Insulated Dual Element Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in X 6.0 ft; 120 VAC
Call For Price
Showing 1- 8 of 8