This product is now discontinued, View Related Products

Brinkmann ChemSaver Dispensers

  • Unique PTFE “wiping” piston seal eliminates reagent residue in the cylinder and prevents “freezing” due to piston swelling and reagent crystallization
  • Safety recirculating valve channels reagent back into bottle during priming
  • Precise graduation scale allowing rapid volume setting
  • Telescopic filling tube adjusts to fit most reagent bottles
  • Dispenser rotates 360 degrees for easy and safe dispensing
  • Fully autoclavable assembly—no need to loosen valve or disassemble
  • Discharge tube surrounded by support sleeve to restrict movement and improve safety
  • Volume adjustment sleeve also rotates 360 degree for maximum visibility at all times
  • Fine volume adjustments for working with viscous or high-density liquids
These dispensers feature a smooth pump-action piston that draws and delivers precise volumes time after time. During delivery, a "wiping" seal of PTFE completely displaces reagent in the cylinder without leaving residue behind. This eliminates "freezing" due to piston swelling and reagent crystallization. Recirculating safety valve channels reagent back into bottle during priming.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Division (mL) Max Volume (mL) Min Volume (mL)
Availability Pricing
Brinkmann 22 22 090-0 ChemSaver Dispenser 0.1-2.5 mL WW-07932-00 Mfr # SNGLS20900
COMPARE
0.05 2.5 0.1
Call For Price
Brinkmann 22 22 100-1 ChemSaver Bottletop Dispenser; 1 to 5 mL WW-07932-01 Mfr # SNGLS21001
COMPARE
0.1 5 1
Call For Price
Brinkmann 22 22 110-8 ChemSaver Bottletop Dispenser; 2 to 10 mL WW-07932-02 Mfr # SNGLS21108
COMPARE
0.2 10 2
Call For Price
Brinkmann 22 22 120-5 ChemSaver Bottletop Dispenser; 5 to 25 mL WW-07932-03 Mfr # SNGLS21205
COMPARE
0.5 25 5
Call For Price
Brinkmann 22 22 130-2 ChemSaver Bottletop Dispenser; 10 to 50 mL WW-07932-04 Mfr # SNGLS21302
COMPARE
1 50 10
Call For Price
Brinkmann 02222140-0 ChemSaver Bottletop Dispenser; 20 to 100 mL WW-07932-05 Mfr # SNGLS21400
COMPARE
1 100 20
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat