BRAND® Macro Manual Pipette Controllers
Superior control to standard pipette bulbs
  • Hydrophobic filter helps prevent overaspiration
  • Fully autoclavable at 121° C
The pipette controllers are simple to operate. Simply squeeze the bellows to charge, lift the lever to aspirate, push down to dispense. Amount of deflection controls speed, from dropwise to full flow. Blow out button provides accurate dispensing of "TC" pipettes. Fits volumetric and serological pipettes from 0.1-200 mL. Four colors help brighten your lab.
22,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Code Max Dispensing (mL) Max Volume (mL)
Availability Pricing
BRAND Macro Pipette Controller, 100 mL, Neutral-Color WW-24550-49 Mfr # 26200
COMPARE
Natural 100 100
₹22,500.00INR / Each
BRAND Macro Pipette Controller, 100 mL, Blue WW-24550-51 Mfr # 26202
COMPARE
Blue 100 100
₹22,500.00INR / Each
BRAND Macro Pipette Controller, 100 mL, Magenta WW-24550-53 Mfr # 26203
COMPARE
Magenta 100 100
₹22,500.00INR / Each