BRAND® BRANDplates® pureGrade™ and pureGrade™ S Untreated Microplates
Medium binding plates for a variety of applications
  • Available nonsterile or sterile for specific applications
  • White (luminescence) black (fluorescence) plates available either solid, or with clear bottom for top or bottom reading
  • Choose from three distinct bottom shapes: round, flat, or C (flat, with curved edge)
  • Offered in 96- and 384-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
These sterile and nonsterile, medium binding, non-treated surface microplates are ideal for covering many standard applications, such as homogenous assays, screening, and storage. The polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Three different bottom shapes are available: round (U) with no edges, flat (F) with a flat bottom, and flat with a curved edge (C) bottom for enhanced mixing.
With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384-well microplates feature rounded well corners which allow optimal uniform meniscus formation.
Strip plates, as well as a handful of convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.
31,300.00 - ₹201,000.00INR / Pkg of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Surface Treatment Well Shape
Availability Pricing
BRANDplates® pureGrade™  Non-treated, Non-Sterile, Microplate, 96-Well, PS, Clear, 350 µL, Standard C-Bottom; 100/PK WW-07903-00 Mfr # 781603
COMPARE
96 pureGrade™ Non-treated Standard C-Bottom
₹31,300.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 96-Well, PS, White, 330 µL, Standard U-Bottom; 100/PK WW-07903-01 Mfr # 781604
COMPARE
96 pureGrade™ Non-treated Standard U-Bottom
₹54,300.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 96-Well, PS, White, 350 µL, Standard F-Bottom; 100/PK WW-07903-05 Mfr # 781605
COMPARE
96 pureGrade™ Non-treated Standard F-Bottom
₹54,300.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 96-Well, PS, Black, 330 µL, Standard U-Bottom; 100/PK WW-07903-06 Mfr # 781607
COMPARE
96 pureGrade™ Non-treated Standard U-Bottom
₹54,300.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 96-Well, PS, Black, 350 µL, Standard F-Bottom; 100/PK WW-07903-07 Mfr # 781608
COMPARE
96 pureGrade™ Non-treated Standard F-Bottom
₹54,300.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 96-Well, PS, White, 330 µL, Clear F-Bottom; 100/PK WW-07903-08 Mfr # 781610
COMPARE
96 pureGrade™ Non-treated Clear F-Bottom
₹155,000.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 96-Well, PS, Black, 330 µL, Clear F-Bottom; 100/PK WW-07903-09 Mfr # 781611
COMPARE
96 pureGrade™ Non-treated Clear F-Bottom
₹155,000.00INR / Pkg of 100
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 384-Well, PWhite, 100 µL, Standard F-Bottom; 50/PK WW-07903-11 Mfr # 781621
COMPARE
384 pureGrade™ Non-treated Standard F-Bottom
₹46,900.00INR / Pkg of 50
BRANDplates® pureGrade™  Non-Treated, Non-Sterile Microplate, 384-Well, PBlack, 100 µL, Standard F-Bottom; 50/PK WW-07903-12 Mfr # 781622
COMPARE
384 pureGrade™ Non-treated Standard F-Bottom
₹46,900.00INR / Pkg of 50
BRANDplates® pureGrade™  Non-Treated, Sterile Microplate, 96-Well, PClear, 350 µL, Standard C-Bottom; 50/PK WW-07903-19 Mfr # 781663
COMPARE
96 pureGrade™ S, Non-treated Standard C-Bottom
₹34,000.00INR / Pkg of 50
BRANDplates® pureGrade™  Non-Treated, Sterile Microplate, 96-Well, PWhite, 350 µL, Standard F-Bottom; 50/PK WW-07903-21 Mfr # 781665
COMPARE
96 pureGrade™ S, Non-treated Standard F-Bottom
₹51,500.00INR / Pkg of 50
BRANDplates® pureGrade™  Non-Treated, Sterile Microplate, 96-Well, PBlack, 350 µL, Standard F-Bottom; 50/PK WW-07903-22 Mfr # 781668
COMPARE
96 pureGrade™ S, Non-treated Standard F-Bottom
₹51,500.00INR / Pkg of 50
BRANDplates® pureGrade™  Non-Treated, Sterile Microplate, 96-Well, PBlack, 330 µL, Clear F-Bottom; 50/PK WW-07903-24 Mfr # 781671
COMPARE
96 pureGrade™ S, Non-treated Clear F-Bottom
₹139,000.00INR / Pkg of 50
BRANDplates® pureGrade™  S, Non-Treated, Sterile Microplate, 384-Well, PS, Clear, 100 µL, Standard F-Bottom; 50/PK WW-07903-26 Mfr # 781680
COMPARE
384 pureGrade™ S, Non-treated Standard F-Bottom
₹46,900.00INR / Pkg of 50
BRANDplates® pureGrade™  Non-Treated, Non-Sterile, Divisible Strip Plate, Clear with Grid, 96-Well, 360 µL, Standard F-Bottom; 100/PK WW-07904-27 Mfr # 782301
COMPARE
96 pureGrade™ Non-treated Standard F-Bottom
₹201,000.00INR / Pkg of 100
Showing 1- 15 of 15