BRAND® Pipette Tips
Pipette tips fit BRAND® Transferpette® and most major brands
  • Available non-sterile or BIO-CERT® sterile
  • Oleamide and diHEMDA-free
  • Rack packed tips up to 1000µL free of DNA, RNase, endotoxin and ATP to detection limit
  • Sterile tips have a Sterility Assurance Level (SAL) of 10-6
BRAND® pipette tips are made of a high-purity polypropylene that is free of additives, lubricants and pigments (bulk 200µL and 1000µL tips feature cadminum-free pigments for easy identification) that cause sample contamination. All tips are manufactured in a clean room under state-of-the-art production conditions and automatically packaged to ensure high quality. Tips can be autoclaved at 121°C (250°F) and 15 psi (1 bar). Choose between non-sterile and BIO-CERT® sterile tips, which adhere to ISO 11137 and AAMI guidelines.
4,500.00 - ₹91,100.00INR / Pkg of 1000
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Sterile
Availability Pricing
BRAND 702600 Bulk Pipette Tips, 0.5-5 mL; 5 Bags, 200/ea WW-24554-93 Mfr # 702600
COMPARE
5000 500 No
₹45,100.00INR / Pkg of 1000
BRAND 702604 Bulk Pipette Tips, 1-10 mL; 5 Bags, 200/ea WW-24554-94 Mfr # 702604
COMPARE
10000 1000 No
₹86,500.00INR / Pkg of 1000
BRAND 702605 Pipette Tips, 0.5-5 mL; 1 Box, 28/ea WW-24554-95 Mfr # 702605
COMPARE
5000 500 No
₹9,900.00INR / Pkg of 28
BRAND 702608 Pipette Tips, 1-10 mL; 1 Box, 18/ea WW-24554-96 Mfr # 702608
COMPARE
10000 1000 No
₹20,100.00INR / Pkg of 18
BRAND 732002 Bulk nano-cap™ Pipette Tips, 0.1-20 uL; 2 Bags, 1000/ea WW-24554-97 Mfr # 732002
COMPARE
20 0.1 No
₹21,900.00INR / Pkg of 2000
BRAND 732008 Bulk Pipette Tips, 2-200 uL, Yellow; 1 Bag, 1000/ea WW-24555-00 Mfr # 732008
COMPARE
200 2 No
₹4,500.00INR / Pkg of 1000
BRAND 732010 Bulk Pipette Tips, 5-300 uL; 2 Bags, 1000/ea WW-24555-01 Mfr # 732010
COMPARE
300 5 No
₹7,000.00INR / Pkg of 1000
BRAND 732012 Bulk Pipette Tips, 50-1000 uL, Blue; 2 Bags, 1000/ea WW-24555-02 Mfr # 732012
COMPARE
1000 50 No
₹4,900.00INR / Pkg of 1000
BRAND 732022 Bulk Pipette Tips, 0.1-20 uL; 10 Bags, 1000/ea WW-24555-03 Mfr # 732022
COMPARE
20 0.1 No
₹91,100.00INR / Pkg of 10000
Showing 1- 9 of 9