This product is now discontinued, View Related Products

Brand Transferpette® Multichannel Pipettors

Calibrate and service quickly and reliably
  • Adjustable handle rotates 360°° for right- or left-hand use
These Transferpette® multichannel pipettors let you fill microplates in a fraction of the time required using single-channel pipettors. Ideal for applications such as immunoassays, antibody testing, and microbial identification.

Adjustable digital setting ensures precise, repeatable volumes and can be recalibrated. Display is located on the handle for convenient viewing even during pipetting. You can easily perform field recalibration without any tools. Simply remove the cover and adjust the volume indicator. This process adapts the pipette to sample conditions and ensures accuracy.

All models accept polypropylene tips (sold separately).
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
BrandTech 2703604 Multichannel pipettors, 8 channel, 2.5-25 µL WW-24550-00 Mfr # 703604
COMPARE
25 2.5 8
Call For Price
BrandTech 2703606 Multichannel pipettors, 8 channel, 10-50 µL WW-24550-02 Mfr # 703606
COMPARE
50 5 8
Call For Price
BrandTech 2703608 Multichannel pipettors, 8 channel, 20-100 µL WW-24550-04 Mfr # 703708
COMPARE
100 10 8
Call For Price
BrandTech 2703610 Multichannel pipettors, 8 channel, 50-200 µL WW-24550-06 Mfr # 703710
COMPARE
200 20 8
Call For Price
BrandTech 2703624 Multichannel pipettors, 12 channel, 5-25 µL WW-24550-10 Mfr # 703724
COMPARE
25 2.5 12
Call For Price
BrandTech 2703626 Transferpette Multichannel pipettors, 12 channel, 10-50 µL WW-24550-12 Mfr # 703726
COMPARE
50 5 12
Call For Price
BrandTech 2703628 Transferpette Multichannel pipettors, 12 channel, 20-100 µL WW-24550-14 Mfr # 703728
COMPARE
100 10 12
Call For Price
BrandTech 2703630 Multichannel pipettors, 12 channel, 20-200µL WW-24550-16 Mfr # 703630
COMPARE
200 20 12
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat