BRAND® Disposable UV-Spectrophotometry Cuvettes
Mold-cavity matched for consistency
  • Suitable for measurements in the UV-spectrum (=230 nm)
  • An excellent choice for molecular Biology applications
  • Expanded polystyrene packaging to preserve optical quality
These cuvettes are made from a unique material that is resistant to aggressive solvents, acids, and bases including chemicals such as acetone, butanone, DMF, and concentrated hydrochloric acid. Suitable for measurements from 230 nm to 900 nm, such as nucleic acid purity measurements. Molded for consistent performance, each cuvette is manufactured from the same mold to ensure the lowest variation in extinction coefficient. 10 mm optical pathlength. Available in macro (2.5 - 4.5 mL); semi-micro (1.5 - 3.0 mL); and ultra-micro (minimum fill volume 70 µL) sizes in both 15 mm and 8.5 mm z-dimensions to fit most major brands of spectrophotometers. Ultra-micro size cuvettes also available individually wrapped.
23,300.00 - ₹107,000.00INR / Pkg of 100
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Capacity (MicroL) Pathlength (mm) Description
Availability Pricing
BRAND Macro Disposable UV-Cuvettes, 2.5 to 4.5 mL; 100/PK WW-06343-71 Mfr # 759170
COMPARE
4500 10 Disposable UV-Cuvettes, Macro 2.5 to 4.5 mL, Bulk Packaged; 100/Pk
₹23,300.00INR / Pkg of 100
BRAND Semi-Micro Disposable UV-Cuvettes, 1.5 to 3 mL; 500/PK WW-06343-72 Mfr # 759165
COMPARE
3000 10 Disposable UV-Cuvettes, Semi-Micro, Bulk Packaged; 500/Pk
₹107,000.00INR / Pkg of 500
BRAND Semi-Micro Disposable UV-Cuvettes, 1.5 to 3 mL; 100/PK WW-06343-73 Mfr # 759150
COMPARE
3000 10 Disposable UV-Cuvettes, Semi-Micro, Bulk Packaged; 100/Pk
₹23,500.00INR / Pkg of 100
BRAND Ultra-Micro Disposable UV-Cuvettes, 15 mm; 500/PK WW-06343-74 Mfr # 759230
COMPARE
550 10 Disposable UV-Cuvettes, Ultra-Micro, 15 mm, Bulk Packaged; 500/Pk
₹107,000.00INR / Pkg of 500
BRAND Ultra-Micro Disposable UV-Cuvettes, 15 mm; 100/PK WW-06343-75 Mfr # 759220
COMPARE
550 10 Disposable UV-Cuvettes, Ultra-Micro, 15 mm, Bulk Packaged; 100/Pk
₹23,300.00INR / Pkg of 100
BRAND Ultra-Micro Disposable UV-Cuvettes, 15 mm; Individually Wrapped; 100/PK WW-06343-76 Mfr # 759235
COMPARE
550 10 Disposable UV-Cuvettes, Ultra-Micro, 15 mm, Individually Wrapped; 100/Pk
₹34,300.00INR / Pkg of 100
BRAND Ultra-Micro Disposable UV-Cuvettes, 8.5 mm; 500/PK WW-06343-77 Mfr # 759210
COMPARE
550 10 Disposable UV-Cuvettes, Ultra-Micro, 8.5 mm, Bulk Packaged; 500/Pk
₹107,000.00INR / Pkg of 500
BRAND Ultra-Micro Disposable UV-Cuvettes, 8.5 mm; 100/PK WW-06343-78 Mfr # 759200
COMPARE
550 10 Disposable UV-Cuvettes, Ultra-Micro, 8.5 mm, Bulk Packaged; 100/Pk
₹23,300.00INR / Pkg of 100
BRAND Ultra-Micro Disposable UV-Cuvettes, 8.5 mm; Individually Wrapped; 100/PK WW-06343-79 Mfr # 759215
COMPARE
550 10 Disposable UV-Cuvettes, Ultra-Micro, 8.5 mm, Individually Wrapped; 100/Pk
₹34,300.00INR / Pkg of 100
Showing 1- 9 of 9