BRAND® Disposable PMMA (Acrylic) Spectrophotometry Cuvettes
Mold-cavity matched for consistency
  • Superior results in the extended visible spectrum (=300 nm)
  • Expanded polystyrene packaging to preserve optical quality
Each and every BRAND cuvette benefits from their over quarter century of experience in precision cuvette molding to the most exacting standards for reproducible results. Cuvettes have 10 mm optical pathlength, and are available in macro (2.5 mL =- 4.5 mL) and semi-micro (1.5 mL - 3.0 mL) sizes.
4,200.00 - ₹22,100.00INR / Pkg of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
BRAND Macro Polymethyl Methacrylate (PMMA, acrylic) Cuvettes; 500/PK WW-06343-55 Mfr # 759080D
COMPARE
Methacrylate 4.5 10
₹22,100.00INR / Pkg of 500
BRAND Macro Polymethyl Methacrylate (PMMA, acrylic) Cuvettes; 100/PK WW-06343-56 Mfr # 759081D
COMPARE
Methacrylate 4.5 10
₹5,100.00INR / Pkg of 100
BRAND Semi-Micro Polymethyl Methacrylate (PMMA, acrylic) Cuvettes; 500/PK WW-06343-57 Mfr # 759085D
COMPARE
Methacrylate 3 10
₹19,100.00INR / Pkg of 500
BRAND Semi-Micro Polymethyl Methacrylate (PMMA, acrylic) Cuvettes; 100/PK WW-06343-58 Mfr # 759086D
COMPARE
Methacrylate 3 10
₹4,200.00INR / Pkg of 100