BNC Adapters
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors. Order an adapter to ensure your meter and electrode are compatible.
7,600.00 - ₹9,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Adapter, BNC meter to U.S. Standard electrode
₹7,600.00INR / Each
COMPARE
Adapter, single pin DIN meter to BNC electrode
₹7,700.00INR / Each
COMPARE
Adapter, U.S. standard meter to BNC electrode
₹9,100.00INR / Each