BLAUBRAND® USP Class A Graduated Glass Cylinders
Meets USP Standards as well as Class A per DIN EN ISO 4788
  • Calibrated "to contain"
  • Hexagonal base
  • Includes USP batch certificate
These BRAND® graduated cylinders comply with the Class A error limits required by the US Pharmacopeia. Each graduated cylinder is calibrated on computer controlled, automated production lines for maximum precision. Full ring marks at major graduations make it easy to read. Features a spout and hexagonal glass base. BLAUBRAND® blue enamel markings are highly legibile, and abrasion resistant. All graduated cylinders come with a batch number and an accompanying batch certificate.
8,800.00 - ₹35,600.00INR / Pkg of 2
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance (mL)
Availability Pricing
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 5 mL; 2/PK WW-06102-01 Mfr # 32805
COMPARE
5 0.1 ±0.05
₹8,800.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 10 mL; 2/PK WW-06102-03 Mfr # 32808
COMPARE
10 0.2 ±0.10
₹8,800.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 25 mL; 2/PK WW-06102-04 Mfr # 32820
COMPARE
25 0.5 ±0.17
₹9,100.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 50 mL; 2/PK WW-06102-05 Mfr # 32828
COMPARE
50 1 ±0.25
₹10,300.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 100 mL; 2/PK WW-06102-06 Mfr # 32838
COMPARE
100 1 ±0.5
₹10,800.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 250 mL; 2/PK WW-06102-07 Mfr # 32848
COMPARE
250 2 ±1.0
₹20,700.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 500 mL; 2/PK WW-06102-08 Mfr # 32854
COMPARE
500 5 ±2.0
₹35,600.00INR / Pkg of 2
BRAND BLAUBRAND® Graduated Cylinder, Glass, Tall Form, Class A, 1000 mL WW-06102-09 Mfr # 32862
COMPARE
1000 10 ±3.0
₹25,900.00INR / Each
Showing 1- 8 of 8