Biological Indictors, Pro Spore Ampoule
ProSpore Ampoule is a self-contained Biological Indicator available for use in steam sterilization. It is ideal for use in validating the sterilization of volumes of liquid that take a longer time to reach sterilizing conditions. The Ampoule is filled with modified Soyabean, Casein digest broth containing bromocresol purple (acid) indicator. Complies with EN 866-7 and AAMI ST 19.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Capacity (cu ft) Max Pressure (PSI) Max Temperature (° F)
Availability Pricing
Pro Spore PA7953105 Ampule Self Contained Steam Bio Indicator 105 Population, 50 Amp/pack WW-94702-30 Mfr # PA7953105
COMPARE
Call For Price
Pro Spore PA7953106 Ampule Self Contained Steam Bio Indicator 106 Population, 50 Amp/pack WW-94702-31 Mfr # PA7953106