SHOWA D-Flex Cut-Resistant Glove
Safely handle glass, sheet and scrap metal, or process food
Lightweight 10-gauge seamless knit glove offers dexterity, tactile sensitivity, and protection. Use to safely handle glass, sheet metal, scrap metal, and to process food. Glove offer superior cut protection with a special wrapping process using Dynema® fibers. Washable and bleachable. Not designed for use with moving or serrated blades. Ambidextrous.
Approval: CE
Compliance: 21CFR, 170-199, acceptable for food and pharmaceutical use.
1,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size
Availability Pricing
SHOWA 910-07 Cut-Resistant Glove, Uncoated; size, small WW-86289-50 Mfr # 910-07
COMPARE
Small
₹1,500.00INR / Each
SHOWA 910-08 Cut-Resistant Glove, Uncoated; size, medium WW-86289-51 Mfr # 910-08
COMPARE
Medium
₹1,500.00INR / Each
SHOWA 910-09 Cut-Resistant Glove, Uncoated; size, large WW-86289-52 Mfr # 910-09
COMPARE
Large
₹1,500.00INR / Each